Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Amino-2-bromobenzoic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

H60382-14 H60382-06 H60382-14 H60382-06
AAH60382-14EA 277.99 USD InStock
CAAAH60382-06 AAH60382-14 AAH60382-06 CAAAH60382-14
5-Amino-2-bromobenzoic acid 95%
5-Amino-2-bromobenzoic acid


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 171…179 °C
Storage Temperature: Ambient
MDL Number: MFCD07368968
CAS Number: 2840-02-0
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty