Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,3,6-Trimethylphenol 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Hydroxypseudocumene

H60489-14 H60489-22 H60489-22 H60489-14
AAH60489-14EA 28.79 USD InStock
AAH60489-14 AAH60489-22 CAAAH60489-14 CAAAH60489-22
2,3,6-Trimethylphenol 95%
2,3,6-Trimethylphenol


Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 215 °C
Melting Pt: 59…62 °C
Density: 0.940
Flash Pt: 100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002229
CAS Number: 2416-94-6
EINECS: 219-330-3
UN: 2430
ADR: "8",III
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty