Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(4-Methoxyphenyl)butyric acid 98%

Supplier: Thermo Scientific Chemicals
Warning

H60613-03 H60613-03 H60613-06 H60613-06
AAH60613-03EA 24.39 USD InStock
CAAAH60613-06 CAAAH60613-03 AAH60613-03 AAH60613-06
4-(4-Methoxyphenyl)butyric acid 98%
4-(4-Methoxyphenyl)butyric acid


Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00004404
CAS Number: 4521-28-2
EINECS: 224-849-3
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty