Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: H-D-Trp-OMe.HClH-D-Trp-OMe·HClD-Tryptophan methyl ester hydrochloride

H60622-03 H60622-06 H60622-03 H60622-06
AAH60622-06EA 80.29 USD InStock
AAH60622-06 AAH60622-03 CAAAH60622-06 CAAAH60622-03
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride


Formula: C₁₂H₁₄N₂O₂·HCl
MW: 254.7 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Ambient
MDL Number: MFCD00038992
CAS Number: 14907-27-8
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty