Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-O-Benzyl-D-glucal 97%

Supplier: Thermo Scientific Chemicals

H60678-MD H60678-03 H60678-03 H60678-MD
AAH60678-MDEA 59.29 USD InStock
CAAAH60678-03 AAH60678-MD CAAAH60678-MD AAH60678-03
4-O-Benzyl-D-glucal 97%
4-O-Benzyl-D-glucal


Formula: C₁₃H₁₆O₄
MW: 236.267 g/mol
Melting Pt: 99…103 °C
Storage Temperature: Ambient
MDL Number: MFCD01863610
CAS Number: 58871-11-7
Order Now
Specification Test Results
Assay Minimum 97

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty