Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) trifluoromethanesulfonate 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Trifluoromethanesulfonic Acid Nickel(II) SaltNickel(II) Triflate

H60878-03 H60878-MD H60878-MD H60878-03
AAH60878-03EA 42.59 USD InStock
AAH60878-03 CAAAH60878-MD AAH60878-MD CAAAH60878-03
Nickel(II) trifluoromethanesulfonate 96%
Nickel(II) trifluoromethanesulfonate


Formula: C₂F₆NiO₆S₂
MW: 356.82 g/mol
Melting Pt: 100…106 °C
Storage Temperature: Ambient
MDL Number: MFCD00673740
CAS Number: 60871-84-3
Order Now
Specification Test Results
Assay Minimum 96

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty