Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,2-Dimethyl-4-pentenoic acid 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 2,2-Dimethylpent-4-enoic acid

H60940-06 H60940-03
AAH60940-03EA 67.59 USD InStock
AAH60940-03 AAH60940-06
2,2-Dimethyl-4-pentenoic acid 95%
2,2-Dimethyl-4-pentenoic acid


Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 104…108 °C
Density: 0.933
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00012304
CAS Number: 16386-93-9
UN: 3265
ADR: "8",III
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty