Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L-Isoserine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-3-Amino-2-hydroxypropionic Acid

H60945-03 H60945-MD H60945-03 H60945-MD
AAH60945-03EA 230.99 USD InStock
AAH60945-03 AAH60945-MD CAAAH60945-03 CAAAH60945-MD
L-Isoserine 98%
L-Isoserine


Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 197…198 °C
Storage Temperature: Ambient
MDL Number: MFCD03428326
CAS Number: 632-13-3
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty