Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methylsuccinic acid 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Pyrotartaric Acid

H60967-22 H60967-14 H60967-14 H60967-22
AAH60967-22EA 319.99 USD InStock
AAH60967-22 CAAAH60967-14 CAAAH60967-22 AAH60967-14
Methylsuccinic acid 99%
Methylsuccinic acid


Formula: C₅H₈O₄
MW: 132.12 g/mol
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00002659
CAS Number: 498-21-5
EINECS: 207-857-1
Order Now
Specification Test Results
Assay Minimum 99

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty