Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Nitro-3,4-dihydro-2H-1,4-benzothiazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H61048-MD H61048-03
AAH61048-MDEA 74.09 USD InStock
AAH61048-MD AAH61048-03
7-Nitro-3,4-dihydro-2H-1,4-benzothiazine 97%
7-Nitro-3,4-dihydro-2H-1,4-benzothiazine


Formula: C₈H₈N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD23379852
CAS Number: 69373-37-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty