Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(4-Cyanophenyl)-2,5-dimethylpyrrole 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile

H61178-06 H61178-03
AAH61178-06EA 320.99 USD InStock
AAH61178-06 AAH61178-03
1-(4-Cyanophenyl)-2,5-dimethylpyrrole 98%
1-(4-Cyanophenyl)-2,5-dimethylpyrrole


Formula: C₁₃H₁₂N₂
MW: 196.25
Storage Temperature: Ambient
MDL Number: MFCD03228548
CAS Number: 119516-86-8
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty