Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-fluorobenzoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H61251-06 H61251-03 H61251-03 H61251-06
AAH61251-06EA 212.99 USD InStock
AAH61251-06 CAAAH61251-03 CAAAH61251-06 AAH61251-03
2-Bromo-6-fluorobenzoic acid 97%
2-Bromo-6-fluorobenzoic acid


Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
Melting Pt: 154…158 °C
Storage Temperature: Ambient
MDL Number: MFCD01569539
CAS Number: 2252-37-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty