Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-3-nitropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

H61260-14 H61260-22 H61260-22 H61260-14
AAH61260-22EA 653.99 USD InStock
CAAAH61260-14 CAAAH61260-22 AAH61260-22 AAH61260-14
2-Fluoro-3-nitropyridine 98%
2-Fluoro-3-nitropyridine


Formula: C₅H₃FN₂O₂
Boiling Pt: 110 °C/10 mm
Melting Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD03095068
CAS Number: 1480-87-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty