Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Carbobenzoxy-D-glutamic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Cbz-D-glutamic AcidZ-D-Glu-OH

H61401-14 H61401-22
AAH61401-14EA 110.99 USD InStock
AAH61401-14 AAH61401-22
N-Carbobenzoxy-D-glutamic acid 95%
N-Carbobenzoxy-D-glutamic acid


Formula: C₁₃H₁₅NO₆
MW: 281.26 g/mol
Melting Pt: 119…121 °C
Storage Temperature: Ambient
MDL Number: MFCD00063193
CAS Number: 63648-73-7
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty