Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Cyano-4-methylaniline 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Amino-2-methylbenzonitrile

H61422-06 H61422-14
AAH61422-14EA 227.99 USD InStock
AAH61422-14 AAH61422-06
3-Cyano-4-methylaniline 97%
3-Cyano-4-methylaniline


Formula: C₈H₈N₂
MW: 132.16 g/mol
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00017608
CAS Number: 50670-64-9
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty