Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Amino-5-bromo-2-chloropyrimidine 95%

Supplier: Thermo Scientific Chemicals
Warning

H61429-06 H61429-14 H61429-14 H61429-03 H61429-06 H61429-03
AAH61429-06EA 142.99 USD InStock
AAH61429-06 AAH61429-14 AAH61429-03 CAAAH61429-06 CAAAH61429-14 CAAAH61429-03
4-Amino-5-bromo-2-chloropyrimidine 95%
4-Amino-5-bromo-2-chloropyrimidine


Formula: C4H3BrClN3
MW: 208.44 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07782040
CAS Number: 205672-25-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty