Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-5-fluoro-2-nitrobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Bromo-4-fluoronitrobenzene2-Bromo-4-fluoro-1-nitrobenzene

H61466-06 H61466-22 H61466-14 H61466-14 H61466-22 H61466-06
AAH61466-06EA 49.69 USD InStock
CAAAH61466-22 AAH61466-06 AAH61466-22 AAH61466-14 CAAAH61466-14 CAAAH61466-06
1-Bromo-5-fluoro-2-nitrobenzene 98%
1-Bromo-5-fluoro-2-nitrobenzene


Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 41…45 °C
Storage Temperature: Ambient
MDL Number: MFCD00792441
CAS Number: 700-36-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty