Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-1,3-dioxolan-2-one 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Fluoro-2-oxo-1,3-dioxolaneFluoroethylene Carbonate

H61502-22 H61502-14 H61502-14 H61502-22
AAH61502-22EA 528.44 USD InStock
CAAAH61502-22 CAAAH61502-14 AAH61502-22 AAH61502-14
4-Fluoro-1,3-dioxolan-2-one 98%
4-Fluoro-1,3-dioxolan-2-one


Formula: C₃H₃FO₃
MW: 106.05 g/mol
Boiling Pt: 212 °C
Melting Pt: 18…23 °C
Flash Pt: >102 °C(216 °F)
Storage Temperature: Ambient
MDL Number: MFCD06247543
CAS Number: 114435-02-8
EINECS: 483-360-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty