Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloromethyl-1H-tetrazole 95%

Supplier: Thermo Scientific Chemicals
Warning

H61643-03 H61643-06 H61643-03 H61643-06 H61643-MD H61643-MD
AAH61643-06EA 353.99 USD InStock
AAH61643-06 CAAAH61643-MD AAH61643-MD CAAAH61643-06 CAAAH61643-03 AAH61643-03
5-Chloromethyl-1H-tetrazole 95%
5-Chloromethyl-1H-tetrazole


Formula: C2H3ClN4
Storage Temperature: Ambient
MDL Number: MFCD00512845
CAS Number: 55408-11-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty