Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl trifluoromethanesulfonate 97%

Supplier: Thermo Scientific Chemicals
Danger

H61758-22 H61758-14
AAH61758-22EA 423.99 USD InStock
AAH61758-22 AAH61758-14
Methyl trifluoromethanesulfonate 97%
Methyl trifluoromethanesulphonate


Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
Boiling Pt: 99 °C
Density: 1.500
Flash Pt: 38 °C(100 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000409
CAS Number: 333-27-7
EINECS: 206-371-7
UN: 2920
ADR: "8",II
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty