Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Fluoro-2-nitroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H61806-14 H61806-22 H61806-06
AAH61806-14EA 77.59 USD InStock
AAH61806-14 AAH61806-22 AAH61806-06
5-Fluoro-2-nitroaniline 97%
5-Fluoro-2-nitroaniline


Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00034065
CAS Number: 2369-11-1
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty