Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

H61858-03 H61858-MD
AAH61858-03EA 308.99 USD InStock
AAH61858-03 AAH61858-MD
2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile 95%
2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile


Formula: C₁₂H₁₄BrN₃
Storage Temperature: Ambient
CAS Number: 1260763-01-6
UN: 3439
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty