Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 97%

Supplier: Thermo Scientific Chemicals
Synonyms: N-(tert-Butoxycarbonyl)-L-glutamic Acid Dimethyl EsterDimethyl N-Boc-L-glutamateN-Boc-L-glutamic Acid Dimethyl EsterBoc-Glu(OMe)-OMe

H62056-06 H62056-14
AAH62056-14EA 304.99 USD InStock
AAH62056-14 AAH62056-06
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate 97%
Dimethyl-N-(tert-butoxycarbonyl)-L-glutamate


Formula: C₁₂H₂₁NO₆
MW: 275.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094774
CAS Number: 59279-60-6
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty