Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-6-fluorobenzylamine hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-(2-Bromo-6-fluorophenyl)methanamine hydrochloride (1:1)Benzenemethanamine, 2-bromo-6-fluoro-, hydrochloride (1:1)

H62086-06 H62086-MD H62086-03
AAH62086-03EA 186.99 USD InStock
AAH62086-03 AAH62086-06 AAH62086-MD
2-Bromo-6-fluorobenzylamine hydrochloride 96%
2-Bromo-6-fluorobenzylamine hydrochloride


Formula: C₇H₇BrFN·HCl
MW: 240.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546112
CAS Number: 1214330-90-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty