Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dimethyl-D-glutamate hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: D-Glutamic Acid Dimethyl Ester HydrochlorideH-D-Glu(OMe)-OMe.HCl

H62132-14 H62132-03 H62132-06
AAH62132-14EA 309.99 USD InStock
AAH62132-14 AAH62132-03 AAH62132-06
Dimethyl-D-glutamate hydrochloride 98%
Dimethyl-D-glutamate hydrochloride


Formula: C₇H₁₃NO₄
MW: 211.64 g/mol
Melting Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00235872
CAS Number: 27025-25-8
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty