Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Z-Sar-OHN-Cbz-N-methylglycineZ-N-Me-Gly-OH

H62135-14 H62135-06
AAH62135-14EA 135.99 USD InStock
AAH62135-14 AAH62135-06
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid 95%
2-(((Benzyloxy)carbonyl)(methyl)amino)acetic acid


Formula: C11H13NO4
MW: 223.3 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00021747
CAS Number: 39608-31-6
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty