Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Asp(OMe)-OH·HClL-Aspartatic Acid 4-Methyl Ester Hydrochloride

H62172-14 H62172-06 H62172-03
AAH62172-14EA 440.67 USD InStock
AAH62172-14 AAH62172-03 AAH62172-06
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 97%
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride


Formula: C5H10ClNO4
MW: 183.6 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038972
CAS Number: 16856-13-6
EINECS: 240-880-5
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty