Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4-fluorobenzylamine hydrochloride 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (2-Bromo-4-fluorophenyl)methanamine hydrochloride

H62200-MD H62200-03 H62200-06
AAH62200-03EA 196.99 USD InStock
AAH62200-03 AAH62200-MD AAH62200-06
2-Bromo-4-fluorobenzylamine hydrochloride 96%
2-Bromo-4-fluorobenzylamine hydrochloride


Formula: C₇H₈BrClFN
MW: 240.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672922
CAS Number: 289038-14-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty