Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-Asp-OBzl 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-D-aspartic acid 1-benzyl ester

H62210-MD H62210-06 H62210-03
AAH62210-03EA 79.59 USD InStock
AAH62210-03 AAH62210-06 AAH62210-MD
Boc-D-Asp-OBzl 98%
Boc-D-Asp-OBzl


Formula: C₁₆H₂₁NO₆
Storage Temperature: Ambient
MDL Number: MFCD00038264
CAS Number: 92828-64-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty