Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-3,5-difluoro-L-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-Phe(3,5-DiF)-OH

H62216-06 H62216-MD H62216-03
AAH62216-03EA 134.99 USD InStock
AAH62216-03 AAH62216-MD AAH62216-06
N-Boc-3,5-difluoro-L-phenylalanine 95%
N-Boc-3,5-difluoro-L-phenylalanine


Formula: C₁₄H₁₇F₂NO₄
MW: 301.29 g/mol
Melting Pt: 115 °C
Storage Temperature: Ambient
MDL Number: MFCD00797555
CAS Number: 205445-52-9
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty