Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-N-tert-Butoxycarbonylalanine methyl ester 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-L-alanine methyl esterBoc-Ala-OMe

H62271-14 H62271-06
AAH62271-14EA 218.99 USD InStock
AAH62271-14 AAH62271-06
(S)-N-tert-Butoxycarbonylalanine methyl ester 95%
(S)-N-tert-Butoxycarbonylalanine methyl ester


Formula: C9H17NO4
MW: 203.24 g/mol
Melting Pt: 32…35 °C
Density: 1.030
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00038513
CAS Number: 28875-17-4
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty