Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Fmoc-L-aspartic AcidFmoc-Asp-OH

H62274-14 H62274-06 H62274-22
AAH62274-06EA 44.89 USD InStock
AAH62274-06 AAH62274-14 AAH62274-22
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid 95%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid


Formula: C₁₉H₁₇NO₆
MW: 355.35 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00237654
CAS Number: 119062-05-4
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty