Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester

Supplier: Thermo Scientific Chemicals
Synonyms: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-benzo[b][1,4]oxazine

H62290-03 H62290-MD
AAH62290-03EA 580.99 USD InStock
AAH62290-03 AAH62290-MD
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester
3,4-Dihydro-2H-1,4-benzoxazine-6-boronic acid pinacol ester


Formula: C₁₄H₂₀BNO₃
MW: 261.128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18073255
CAS Number: 1155264-46-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty