Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Fmoc-S-trityl-D-cysteineFmoc-D-Cys(Trt)-OH

H62301-06 H62301-03
AAH62301-06EA 79.49 USD InStock
AAH62301-06 AAH62301-03
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(triphenylmethyl)-D-cysteine


Formula: C₃₇H₃₁NO₄S
MW: 585.72 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00151922
CAS Number: 167015-11-4
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty