Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-DL-phenylglycine 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-DL-Phg-OH

H62354-06 H62354-14
AAH62354-06EA 61.79 USD InStock
AAH62354-06 AAH62354-14
N-Boc-DL-phenylglycine 98%
N-Boc-DL-phenylglycine


Formula: C₁₃H₁₇NO₄
MW: 251.282 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00971063
CAS Number: 3601-66-9
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty