Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-L-Pro-OMe 96%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-L-proline methyl esterN-(tert-Butoxycarbonyl)-L-proline methyll esterBoc-Pro-OMe

H62363-14 H62363-22
AAH62363-14EA 76.29 USD InStock
AAH62363-14 AAH62363-22
Boc-L-Pro-OMe 96%
Boc-L-Pro-OMe


Formula: C11H19NO4
MW: 229.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01075092
CAS Number: 59936-29-7
Order Now
Specification Test Results
Assay 96%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty