Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Alanine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Methyl (R)-2-Aminopropionate Hydrochloride(R)-2-Aminopropionic Acid Methyl Ester HydrochlorideH-D-Ala-OMe.HCl

H62472-14 H62472-06 H62472-22
AAH62472-14EA 129.99 USD InStock
AAH62472-14 AAH62472-22 AAH62472-06
D-Alanine methyl ester hydrochloride 98%
D-Alanine methyl ester hydrochloride


Formula: C₄H₉NO₂
MW: 139.58 g/mol
Melting Pt: 108…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00066141
CAS Number: 14316-06-4
EINECS: 238-258-3
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty