Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-4-fluoro-2-methylaniline 96%

Supplier: Thermo Scientific Chemicals
Warning

H62480-03 H62480-06
AAH62480-03EA 53.25 USD InStock
AAH62480-03 AAH62480-06
5-Bromo-4-fluoro-2-methylaniline 96%
5-Bromo-4-fluoro-2-methylaniline


Formula: C₇H₇BrFN
MW: 204.04 g/mol
Melting Pt: 86…90 °C
Storage Temperature: Ambient
MDL Number: MFCD05865218
CAS Number: 627871-16-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty