Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-His-OMe 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-L-histidine methyl ester

H62545-06 H62545-MD H62545-03
AAH62545-MDEA 28.27 USD InStock
AAH62545-MD AAH62545-06 AAH62545-03
Boc-His-OMe 95%
Boc-His-OMe


Formula: C₁₂H₁₉N₃O₄
Storage Temperature: Ambient
MDL Number: MFCD00167635
CAS Number: 2488-14-4
Order Now
Specification Test Results
Assay Minimum 95

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty