Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-Asp-OMe 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-L-aspartic acid 1-Methyl esterN-(tert-Butoxycarbonyl)-L-aspartic acid 1-Methyl ester1-Methyl N-(tert-Butoxycarbonyl)-L-aspartateN-Boc-L-aspartic acid 1-methyl esterBoc-Asp-OMe1-Methyl N-Boc-L-aspartate

H62590-MD H62590-06 H62590-03
AAH62590-MDEA 60.59 USD InStock
AAH62590-MD AAH62590-06 AAH62590-03
Boc-Asp-OMe 98%
Boc-Asp-OMe


Formula: C₁₀H₁₇NO₆
MW: 247.247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094777
CAS Number: 98045-03-5
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty