Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-D-Lys(Boc)-OH 98%

Supplier: Thermo Scientific Chemicals
Synonyms: Nepsilon-Boc-D-lysine

H62736-03 H62736-MD
AAH62736-MDEA 45.74 USD InStock
AAH62736-MD AAH62736-03
H-D-Lys(Boc)-OH 98%
H-D-Lys(Boc)-OH


Formula: C₁₁H₂₂N₂O₄
MW: 246.307 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00076958
CAS Number: 31202-69-4
Order Now
Specification Test Results
Assay Minimum 98

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty