Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-(tert-Butoxycarbonyl)-D-glutamic acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Boc-D-glutamic AcidBoc-D-Glu-OH

H62762-06 H62762-14 H62762-03
AAH62762-14EA 410.99 USD InStock
AAH62762-14 AAH62762-03 AAH62762-06
N-(tert-Butoxycarbonyl)-D-glutamic acid 98%
N-(tert-Butoxycarbonyl)-D-glutamic acid


Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00190790
CAS Number: 34404-28-9
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty