Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Orn(boc)-OH 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (S)-2-Amino-5-((tert-butoxycarbonyl)amino)pentanoic acidN-α-t-Boc-L-Ornithine MonohydrateNdelta-Boc-L-ornithineBoc-L-Orn-OH

H62774-03 H62774-06
AAH62774-06EA 200.99 USD InStock
AAH62774-06 AAH62774-03
H-Orn(boc)-OH 98%
H-Orn(boc)-OH


Formula: C10H20N2O4
MW: 232.28 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00037219
CAS Number: 13650-49-2
Order Now
Specification Test Results
Assay 98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty