Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-2-cyclopentylacetic acid 95%

Supplier: Thermo Scientific Chemicals
Synonyms: L-CyclopentylglycineH-Cyclopentyl-Gly-OH

H62776-06 H62776-MD H62776-03
AAH62776-03EA 258.99 USD InStock
AAH62776-03 AAH62776-06 AAH62776-MD
(S)-2-Amino-2-cyclopentylacetic acid 95%
(S)-2-Amino-2-cyclopentylacetic acid


Formula: C7H13NO2
MW: 143.18 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788076
CAS Number: 2521-84-8
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty