Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

potassium trimethylacetate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Potassium trimethylacetatePivalic acid potassium salt2,2-Dimethylpropionic acid potassium saltPotassium PivalateTrimethylacetic acid potassium saltPotassium 2,2-Dimethylpropionate

H62803-14 H62803-22 H62803-14 H62803-22
AAH62803-14EA 47.79 USD InStock
AAH62803-14 CAAAH62803-14 CAAAH62803-22 AAH62803-22
potassium trimethylacetate 95%
potassium trimethylacetate


Formula: C₅H₉KO₂
MW: 140.223 g/mol
Melting Pt: ca 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00671345
CAS Number: 19455-23-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty