Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Boc-2-bromo-D-phenylalanine 95%

Supplier: Thermo Scientific Chemicals
Synonyms: Boc-D-Phe(2-Br)-OH

H62848-MD H62848-06 H62848-03
AAH62848-06EA 542.99 USD InStock
AAH62848-06 AAH62848-03 AAH62848-MD
N-Boc-2-bromo-D-phenylalanine 95%
N-Boc-2-bromo-D-phenylalanine


Formula: C₁₄H₁₈BrNO₄
MW: 344.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01317703
CAS Number: 261360-76-3
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty