Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: H-Phg-OMe·HCl(S)-Methyl 2-amino-2-phenylacetate hydrochloride

H62874-22 H62874-14
AAH62874-22EA 176.99 USD InStock
AAH62874-22 AAH62874-14
(S)-(+)-2-Phenylglycine methyl ester hydrochloride 97%
(S)-(+)-2-Phenylglycine methyl ester hydrochloride


Formula: C₉H₁₂ClNO₂
MW: 201.65 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00077158
CAS Number: 15028-39-4
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty