Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-[cis-4-(Boc-amino)cyclohexyl]ethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

H62917-03 H62917-MD
AAH62917-MDEA 107.99 USD InStock
AAH62917-MD AAH62917-03
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine 97%
2-[cis-4-(Boc-amino)cyclohexyl]ethylamine


Formula: C₁₃H₂₆N₂O₂
MW: 242.363 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03844601
CAS Number: 509143-12-8
Order Now
Specification Test Results
Assay 97%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty