Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Benzyl-N-carbobenzoxy-L-glutamate 95%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester1-Benzyl N-Cbz-L-glutamateN-Cbz-L-glutamic Acid 1-Benzyl EsterZ-Glu-OBzl

H62975-14 H62975-06
AAH62975-06EA 47.89 USD InStock
AAH62975-06 AAH62975-14
1-Benzyl-N-carbobenzoxy-L-glutamate 95%
1-Benzyl-N-carbobenzoxy-L-glutamate


Formula: C₂₀H₂₁NO₆
MW: 371.39 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00077013
CAS Number: 3705-42-8
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty