Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-Lys(boc)-OtBu·HCl 95%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Lys(Boc)-OtBu·HCl

H62993-MD H62993-03 H62993-06
AAH62993-06EA 414.99 USD InStock
AAH62993-06 AAH62993-MD AAH62993-03
H-Lys(boc)-OtBu·HCl 95%
H-Lys(boc)-OtBu·HCl


Formula: C₁₅H₃₁ClN₂O₄
MW: 338.873 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00038899
CAS Number: 13288-57-8
Order Now
Specification Test Results
Assay 95%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty